5-Amino-2-chloro-4-methylpyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-methylpyrimidin-5-amine

076428-250MG 076428-1G 076428-5G
102546-130EA 684.62 USD
102546-130 102546-132 102546-134
5-Amino-2-chloro-4-methylpyrimidine ≥95%
5-Amino-2-chloro-4-methylpyrimidine

Hazard: Irritant

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
MDL Number: MFCD12923128
CAS Number: 20090-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR