1-(6-Fluoropyridin-3-yl)ethanone ≥95%
Supplier: Matrix Scientific
076427-250MG
076427-1G
076427-5G
102546-124EA
640
USD
102546-124
102546-126
102546-128
1-(6-Fluoropyridin-3-yl)ethanone ≥95%
1-(6-Fluoropyridin-3-yl)ethanone
Hazard: Irritant
Formula:
C₇H₆FNO MW: 139.1300 g/mol |
MDL Number:
MFCD07375076 CAS Number: 84331-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...