Ethyl-5-bromo-2-phenylthiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-bromo-2-phenylthiazole-4-carboxylate

076423-1G 076423-5G
102546-108EA 640 USD
102546-108 102546-110
Ethyl-5-bromo-2-phenylthiazole-4-carboxylate ≥95%
Ethyl-5-bromo-2-phenylthiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀BrNO₂S
MW: 312.19 g/mol
MDL Number: MFCD08275710
CAS Number: 914347-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR