(S)-4,4-Difluoropyrrolidine-2-carboxylic acid-2,2,2-trifluoroacetic acid ≥95%
Supplier: Matrix Scientific
076422-250MG
076422-1G
102546-104EA
967.69
USD
102546-104
102546-106
(S)-4,4-Difluoropyrrolidine-2-carboxylic acid-2,2,2-trifluoroacetic acid ≥95%
(S)-4,4-Difluoropyrrolidine-2-carboxylic acid-2,2,2-trifluoroacetic acid
Hazard: Irritant
Formula:
C₇H₈F₅NO₄ |
MDL Number:
MFCD22194317 CAS Number: 1373255-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...