3-(9H-Carbazol-9-yl)phenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(9-Carbazolyl)benzeneboronic acid, 3-(9H-Carbazol-9-yl)benzeneboronic Acid, (3-(9H-Carbazol-9-yl)phenyl)boronic acid

076421-1G 076421-5G
102546-100EA 44.62 USD
102546-100 102546-102
3-(9H-Carbazol-9-yl)phenylboronic acid ≥95%
3-(9H-Carbazol-9-yl)phenylboronic acid

Hazard: Irritant

Formula: C₁₈H₁₄BNO₂
MW: 287.13 g/mol
MDL Number: MFCD18072506
CAS Number: 864377-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR