(R)-3-Phenylpyrrolidine hydrochloride ≥95%

Supplier: Matrix Scientific

076420-250MG 076420-1G 076420-5G
102546-094EA 498.46 USD
102546-094 102546-096 102546-098
(R)-3-Phenylpyrrolidine hydrochloride ≥95%
(R)-3-Phenylpyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClN
MDL Number: MFCD06796636
CAS Number: 851000-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR