2-Bromoanthracene ≥95%

Supplier: Matrix Scientific

076417-1G 076417-5G
102546-078EA 710.77 USD
102546-078 102546-080
2-Bromoanthracene ≥95%
2-Bromoanthracene

Hazard: Irritant

Formula: C₁₄H₉Br
MW: 257.1300 g/mol
MDL Number: MFCD07784002
CAS Number: 7321-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR