3-Chloro-5-hydroxybenzaldehyde ≥95%

Supplier: Matrix Scientific

076415-1G 076415-5G
102546-068EA 136.92 USD
102546-068 102546-070
3-Chloro-5-hydroxybenzaldehyde ≥95%
3-Chloro-5-hydroxybenzaldehyde

Hazard: Irritant

Formula: C₇H₅ClO₂
MW: 156.5664 g/mol
MDL Number: MFCD08234659
CAS Number: 1829-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR