Methyl-4-(bromomethyl)-3-chlorobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-(bromomethyl)-3-chlorobenzoate

076414-250MG 076414-1G 076414-5G
102546-060EA 302.08 USD
102546-060 102546-062 102546-064
Methyl-4-(bromomethyl)-3-chlorobenzoate ≥95%
Methyl-4-(bromomethyl)-3-chlorobenzoate

Hazard: Irritant

Formula: C₉H₈BrClO₂
MW: 263.52 g/mol
MDL Number: MFCD10566565
CAS Number: 74733-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR