1H-Indazole-5-sulfonyl chloride ≥95%

Supplier: Matrix Scientific

076413-250MG 076413-1G 076413-5G
102546-054EA 600 USD
102546-054 102546-056 102546-058
1H-Indazole-5-sulfonyl chloride ≥95%
1H-Indazole-5-sulfonyl chloride

Hazard: Corrosive

Formula: C₇H₅ClN₂O₂S
MW: 216.65 g/mol
MDL Number: MFCD11650650
CAS Number: 599183-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR