3-(3-Bromophenyl)acrylaldehyde ≥95%

Supplier: Matrix Scientific

076412-250MG 076412-1G 076412-5G
102546-048EA 400 USD
102546-048 102546-050 102546-052
3-(3-Bromophenyl)acrylaldehyde ≥95%
3-(3-Bromophenyl)acrylaldehyde

Hazard: Irritant

Formula: C₉H₇BrO
MDL Number: MFCD00996500
CAS Number: 15185-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR