Methyl-5-methylthiazole-2-carboxylate ≥95%
Supplier: Matrix Scientific
076411-250MG
076411-1G
076411-5G
102546-042EA
355.38
USD
102546-042
102546-044
102546-046
Methyl-5-methylthiazole-2-carboxylate ≥95%
Methyl-5-methylthiazole-2-carboxylate
Hazard: Irritant
Formula:
C₆H₇NO₂S |
MDL Number:
MFCD01924773 CAS Number: 79247-98-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...