Methyl-2-cyclopentyl-2-hydroxy-2-phenylacetate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-cyclopentyl-2-hydroxy-2-phenylacetate

076408-1G 076408-5G 076408-10G
102546-026EA 55.38 USD
102546-026 102546-028 102546-030
Methyl-2-cyclopentyl-2-hydroxy-2-phenylacetate ≥95%
Methyl-2-cyclopentyl-2-hydroxy-2-phenylacetate

Hazard: Irritant

Formula: C₁₄H₁₈O₃
MW: 234.2900 g/mol
MDL Number: MFCD00019295
CAS Number: 19833-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR