(±)-4-Amino-2-hydroxybutyric acid ≥95%

Supplier: Matrix Scientific

076405-1G 076405-5G 076405-10G
102546-008EA 50.77 USD
102546-008 102546-010 102546-012
(±)-4-Amino-2-hydroxybutyric acid ≥95%
(±)-4-Amino-2-hydroxybutyric Acid

Hazard: Irritant

Formula: C₄H₉NO₃
MDL Number: MFCD00797835
CAS Number: 13477-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR