(R)-1-Methoxypropan-2-amine hydrochloride ≥95%

Supplier: Matrix Scientific

076402-250MG 076402-1G 076402-5G
102545-992EA 800 USD
102545-992 102545-994 102545-996
(R)-1-Methoxypropan-2-amine hydrochloride ≥95%
(R)-1-Methoxypropan-2-amine hydrochloride

Hazard: Irritant

Formula: C₄H₁₂ClNO
MDL Number: MFCD12405301
CAS Number: 626220-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR