(S)-3-Hydroxy-γ-butyrolactone ≥85%

Supplier: Matrix Scientific

Synonyms: (S)-4-Hydroxydihydrofuran-2(3H)-one, (S)-4-Hydroxytetrahydrofuran-2-one

076401-1G 076401-5G 076401-25G
102545-986EA 78.46 USD
102545-986 102545-988 102833-922
(S)-3-Hydroxy-γ-butyrolactone ≥85%
(S)-3-Hydroxy-gamma-butyrolactone

Hazard: Irritant

Formula: C₄H₆O₃
MW: 102.09 g/mol
MDL Number: MFCD00211247
CAS Number: 7331-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR