Ethyl-2-oxazolecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl oxazole-2-carboxylate

076400-250MG 076400-1G
102545-982EA 684.62 USD
102545-982 102545-984
Ethyl-2-oxazolecarboxylate ≥95%
Ethyl 2-oxazolecarboxylate

Hazard: Irritant

Formula: C₆H₇NO₃
MW: 141.1300 g/mol
MDL Number: MFCD04114939
CAS Number: 33036-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR