2-Chloro-6-fluorobenzo[d]oxazole ≥95%

Supplier: Matrix Scientific

076399-250MG 076399-1G 076399-5G
102545-976EA 684.62 USD
102545-976 102545-978 102545-980
2-Chloro-6-fluorobenzo[d]oxazole ≥95%
2-Chloro-6-fluorobenzo[d]oxazole

Hazard: Irritant

Formula: C₇H₃ClFNO
MW: 171.56 g/mol
MDL Number: MFCD16250625
CAS Number: 153403-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR