3,5-Dichloropyridin-4-ol ≥95%

Supplier: Matrix Scientific

076397-250MG 076397-1G 076397-5G
102545-964EA 243.08 USD
102545-964 102545-966 102545-968
3,5-Dichloropyridin-4-ol ≥95%
3,5-Dichloropyridin-4-ol

Hazard: Irritant

Formula: C5H3Cl2NO
MDL Number: MFCD00047448
CAS Number: 17228-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR