Cyclobutylhydrazine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Cyclobutylhydrazine hydrochloride

076395-250MG 076395-1G 076395-5G
102545-954EA 355.38 USD
102545-954 102545-956 102545-958
Cyclobutylhydrazine hydrochloride ≥95%
Cyclobutylhydrazine hydrochloride

Hazard: Irritant

Formula: C₄H₁₁ClN₂
MW: 122.6 g/mol
MDL Number: MFCD06658415
CAS Number: 158001-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR