(S)-Propane-1,2-diamine dihydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
(S)-1,2-Diaminopropane dihydrochloride, (S)-(-)-1,2-Diaminopropane dihydrochloride, S-(-)-Propylenediamine dihydrochloride, (S)-Propylenediamine dihydrochloride, (S)-1,2-Propanediamine dihydrochloride
076394-1G
076394-5G
076394-10G
102545-948EA
21.54
USD
102545-948
102545-950
102545-952
(S)-Propane-1,2-diamine dihydrochloride ≥95%
(S)-Propane-1,2-diamine dihydrochloride
Hazard: Irritant
Formula:
C₃H₁₂Cl₂N₂ MW: 147.05 g/mol |
MDL Number:
MFCD00050706 CAS Number: 19777-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...