tert-Butyl thiazol-4-ylcarbamate ≥95%

Supplier: Matrix Scientific

076393-250MG 076393-1G 076393-5G
102545-942EA 355.38 USD
102545-942 102545-944 102545-946
tert-Butyl thiazol-4-ylcarbamate ≥95%
tert-Butyl thiazol-4-ylcarbamate

Hazard: Irritant

Formula: C₈H₁₂N₂O₂S
MW: 200.2600 g/mol
MDL Number: MFCD18800853
CAS Number: 1235406-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR