Thiophene-2,3-dicarboxylic acid ≥95%

Supplier: Matrix Scientific

076389-250MG 076389-1G
102545-920EA 146.15 USD
102545-920 102545-922
Thiophene-2,3-dicarboxylic acid ≥95%
Thiophene-2,3-dicarboxylic acid

Hazard: Irritant

Formula: C₆H₄O₄S
MDL Number: MFCD00034738
CAS Number: 1451-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR