4-Fluoroisoindoline hydrochloride ≥95%

Supplier: Matrix Scientific

076386-250MG 076386-1G 076386-5G
102545-904EA 1298.46 USD
102545-904 102545-906 102545-908
4-Fluoroisoindoline hydrochloride ≥95%
4-Fluoroisoindoline hydrochloride

Hazard: Irritant

Formula: C₈H₉ClFN
MW: 173.6152 g/mol
MDL Number: MFCD10700143
CAS Number: 924305-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR