(S)-β-(Carbobenzoxyamino)-γ-butyrolactone ≥95%

Supplier: Matrix Scientific

076383-1G 076383-5G 076383-10G
102545-886EA 213.85 USD
102545-886 102545-888 102545-890
(S)-β-(Carbobenzoxyamino)-γ-butyrolactone ≥95%
(S)-beta-(Carbobenzoxyamino)-gamma-butyrolactone

Hazard: Irritant

Formula: C₁₂H₁₃NO₄
MDL Number: MFCD00216569
CAS Number: 87219-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR