2-(Methoxycarbonyl)-4-pyridineboronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate

076379-250MG 076379-1G
102545-868EA 967.69 USD
102545-868 102545-870
2-(Methoxycarbonyl)-4-pyridineboronic acid pinacol ester ≥95%
2-(Methoxycarbonyl)-4-pyridineboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₃H₁₈BNO₄
MDL Number: MFCD08752647
CAS Number: 957062-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR