Ethyl-2-oxo-2-(pyridin-3-yl)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-oxo-2-(pyridin-3-yl)acetate

076378-250MG 076378-1G 076378-5G
102545-862EA 498.46 USD
102545-862 102545-864 102545-866
Ethyl-2-oxo-2-(pyridin-3-yl)acetate ≥95%
Ethyl-2-oxo-2-(pyridin-3-yl)acetate

Hazard: Irritant

Formula: C₉H₉NO₃
MW: 179.1727 g/mol
MDL Number: MFCD09842445
CAS Number: 73672-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR