5-Bromo-2-methox-3-methylypyridine ≥95%
Supplier: Matrix Scientific
076376-1G
076376-5G
076376-10G
102545-850EA
284.62
USD
102545-850
102545-852
102545-854
5-Bromo-2-methox-3-methylypyridine ≥95%
5-Bromo-2-methox-3-methylypyridine
Hazard: Irritant
Formula:
C₇H₈BrNO |
MDL Number:
MFCD07375124 CAS Number: 760207-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...