1-Boc-3-(bromomethyl)piperidine ≥95%

Supplier: Matrix Scientific

Synonyms: N-Boc-3-(bromomethyl)piperidine, 3-(Bromomethyl)piperidine, N-BOC protected

076375-1G 076375-5G 076375-10G
102545-844EA 153.85 USD
102545-844 102545-846 102545-848
1-Boc-3-(bromomethyl)piperidine ≥95%
1-Boc-3-(bromomethyl)piperidine

Hazard: Irritant

Formula: C₁₁H₂₀BrNO₂
MDL Number: MFCD04115539
CAS Number: 193629-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR