2,6-Dichloro-3-methylpyridine ≥95%

Supplier: Matrix Scientific

076372-1G 076372-5G
102545-830EA 44.62 USD
102545-830 102545-832
2,6-Dichloro-3-methylpyridine ≥95%
2,6-Dichloro-3-methylpyridine

Hazard: Irritant

Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Melting Pt: 54…56 °C
MDL Number: MFCD00137847
CAS Number: 58584-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR