2-Amino-4-methoxybenzonitrile ≥95%
Supplier: Matrix Scientific
076371-1G
076371-5G
102545-824EA
347.69
USD
102545-824
102545-826
2-Amino-4-methoxybenzonitrile ≥95%
2-Amino-4-methoxybenzonitrile
Hazard: Irritant
Formula:
C₈H₈N₂O MW: 148.1600 g/mol |
MDL Number:
MFCD05149246 CAS Number: 38487-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...