4-Hydroxy-2,6-dimethoxybenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Dimethoxy-4-hydroxybenzaldehyde

076370-1G 076370-5G 076370-10G
102545-818EA 143.08 USD
102545-818 102545-820 102545-822
4-Hydroxy-2,6-dimethoxybenzaldehyde ≥95%
4-Hydroxy-2,6-dimethoxybenzaldehyde

Hazard: Irritant

Formula: C₉H₁₀O₄
MW: 182.1733 g/mol
MDL Number: MFCD01407645
CAS Number: 22080-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR