2,6-Dichloropyridine-3,4-diamine ≥95%
Supplier: Matrix Scientific
Synonyms:
3,4-Diamino-2,6-dichloropyridine
076366-250MG
076366-1G
076366-5G
102545-794EA
364.62
USD
102545-794
102545-796
102545-798
2,6-Dichloropyridine-3,4-diamine ≥95%
2,6-Dichloropyridine-3,4-diamine
Hazard: Irritant
Formula:
C₅H₅Cl₂N₃ MW: 178.0200 g/mol |
MDL Number:
MFCD10697689 CAS Number: 101079-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...