3-Ethyl-1-phenyl-1H-pyrazol-5-amine ≥95%

Supplier: Matrix Scientific

076364-250MG 076364-1G 076364-5G
102545-782EA 826.15 USD
102545-782 102545-784 102545-786
3-Ethyl-1-phenyl-1H-pyrazol-5-amine ≥95%
3-Ethyl-1-phenyl-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C11H13N3
MW: 187.2400 g/mol
MDL Number: MFCD02255835
CAS Number: 1017781-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR