3-Chloro-4-hydroxypyridin-2(1H)-one ≥95%

Supplier: Matrix Scientific

076362-1G 076362-5G 076362-10G
102545-770EA 246.15 USD
102545-770 102545-772 102545-774
3-Chloro-4-hydroxypyridin-2(1H)-one ≥95%
3-Chloro-4-hydroxypyridin-2(1H)-one

Hazard: Irritant

Formula: C5H4ClNO2
CAS Number: 103792-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR