2-(4-Bromophenyl)pyrrolidine ≥95%

Supplier: Matrix Scientific

076361-1G 076361-5G
102545-766EA 221.54 USD
102545-766 102545-768
2-(4-Bromophenyl)pyrrolidine ≥95%
2-(4-Bromophenyl)pyrrolidine

Hazard: Irritant

Formula: C₁₀H₁₂BrN
MW: 226.1130 g/mol
MDL Number: MFCD02663432
CAS Number: 383127-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR