2-Bromo-5-phenylpyridine ≥95%

Supplier: Matrix Scientific

076359-250MG 076359-1G
102545-758EA 569.23 USD
102545-758 102545-760
2-Bromo-5-phenylpyridine ≥95%
2-Bromo-5-phenylpyridine

Hazard: Irritant

Formula: C₁₁H₈BrN
MDL Number: MFCD04114195
CAS Number: 107351-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR