Ethyl-6-phenylimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

076358-250MG 076358-1G 076358-5G
102545-752EA 640 USD
102545-752 102545-754 102545-756
Ethyl-6-phenylimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 6-phenylimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₆H₁₄N₂O₂
MDL Number: MFCD11111540
CAS Number: 1173694-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR