4-Chloropiperidine hydrochloride ≥95%

Supplier: Matrix Scientific

076355-1G 076355-5G 076355-10G
102545-734EA 427.69 USD
102545-734 102545-736 102545-738
4-Chloropiperidine hydrochloride ≥95%
4-Chloropiperidine hydrochloride

Hazard: Irritant

Formula: C₅H₁₁Cl₂N
MW: 156.0500 g/mol
MDL Number: MFCD03411587
CAS Number: 5382-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR