6-Chloro-5-methylpyridine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

076354-250MG 076354-1G 076354-5G
102545-728EA 456.92 USD
102545-728 102545-730 102545-732
6-Chloro-5-methylpyridine-2-carboxylic acid ≥95%
6-Chloro-5-methylpyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₆ClNO₂
MDL Number: MFCD13176716
CAS Number: 1166828-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR