2-Bromo-6-iodobenzonitrile ≥95%
Supplier: Matrix Scientific
076353-1G
076353-5G
102545-724EA
84.62
USD
102545-724
102545-726
2-Bromo-6-iodobenzonitrile ≥95%
2-Bromo-6-iodobenzonitrile
Hazard: Irritant
Formula:
C₇H₃BrIN MW: 307.9139 g/mol |
MDL Number:
MFCD16619725 CAS Number: 1245648-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...