2-Bromo-5-methylbenzaldehyde ≥95%

Supplier: Matrix Scientific

076351-1G 076351-5G 076351-25G
102545-710EA 75.38 USD
102545-710 102545-712 102833-920
2-Bromo-5-methylbenzaldehyde ≥95%
2-Bromo-5-methylbenzaldehyde

Hazard: Irritant

Formula: C₈H₇BrO
MDL Number: MFCD08669550
CAS Number: 90221-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR