1-Methyl-2-pyrrolecarboxaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: N-Methylpyrrole-2-carboxaldehyde, 1-Methylpyrrole-2-aldehyde, 1-Methylpyrrole-2-carboxaldehyde, 1-Methylpyrrol-2-carbaldehyde, 2-Formyl-1-methylpyrrole

076350-5G 076350-10G 076350-100G
102545-704EA 107.69 USD
102545-704 102545-706 102545-708
1-Methyl-2-pyrrolecarboxaldehyde ≥95%
1-Methyl-2-pyrrolecarboxaldehyde

Hazard: Irritant

Formula: C₆H₇NO
MW: 109.13 g/mol
MDL Number: MFCD00003087
CAS Number: 1192-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR