1-Bromo-4-methoxybutane ≥95%

Supplier: Matrix Scientific

076345-5G 076345-100G
102545-682EA 29.23 USD
102545-682 102545-686
1-Bromo-4-methoxybutane ≥95%
1-Bromo-4-methoxybutane

Hazard: Irritant

Formula: C₅H₁₁BrO
MDL Number: MFCD00037102
CAS Number: 4457-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR