6-(Methylthio)-1H-pyrazolo[3,4-d]pyrimidine ≥95%

Supplier: Matrix Scientific

076343-250MG 076343-1G
102545-674EA 320 USD
102545-674 102545-676
6-(Methylthio)-1H-pyrazolo[3,4-d]pyrimidine ≥95%
6-(Methylthio)-1H-pyrazolo[3,4-d]pyrimidine

Hazard: Irritant

Formula: C₆H₆N₄S
MDL Number: MFCD11040464
CAS Number: 55084-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR