Piperidine-4-carbonitrile hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Cyanopiperidine hydrochloride

076340-1G 076340-5G 076340-10G
102545-658EA 178.46 USD
102545-658 102545-660 102545-662
Piperidine-4-carbonitrile hydrochloride ≥95%
Piperidine-4-carbonitrile hydrochloride

Hazard: Irritant

Formula: C₆H₁₁ClN₂
MW: 146.62 g/mol
MDL Number: MFCD09037930
CAS Number: 240401-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR