((2-(6-Amino-9H-purin-9-yl)ethoxy)methyl)phosphonic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Adefovir

076338-1G 076338-5G
102545-648EA 178.46 USD
102545-648 102545-650
((2-(6-Amino-9H-purin-9-yl)ethoxy)methyl)phosphonic acid ≥95%
((2-(6-Amino-9H-purin-9-yl)ethoxy)methyl)phosphonic acid

Hazard: Irritant

Formula: C₈H₁₂N₅O₄P
MW: 273.19 g/mol
MDL Number: MFCD00866943
CAS Number: 106941-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR