3-Chloro-4-((3-fluorobenzyl)oxy)aniline ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Chloro-4-(3-fluorobenzyloxy)aniline
076336-1G
076336-5G
102545-640EA
46.15
USD
102545-640
102545-642
3-Chloro-4-((3-fluorobenzyl)oxy)aniline ≥95%
3-Chloro-4-((3-fluorobenzyl)oxy)aniline
Hazard: Irritant
Formula:
C₁₃H₁₁ClFNO MW: 251.69 g/mol |
MDL Number:
MFCD06809822 CAS Number: 202197-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...