Candesartan cilexetil ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(((Cyclohexyloxy)carbonyl)oxy)ethyl 1-((2'-(2H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate

076335-1G 076335-5G
102545-636EA 233.85 USD
102545-636 102545-638
Candesartan cilexetil ≥95%
Candesartan cilexetil

Hazard: Irritant

Formula: C₃₃H₃₄N₆O₆
MW: 610.67 g/mol
MDL Number: MFCD00871371
CAS Number: 145040-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR