(2-Fluorophenyl)(2-(methylamino)-5-nitrophenyl)methanone ≥95%

Supplier: Matrix Scientific

076334-1G 076334-5G 076334-10G
102545-630EA 427.69 USD
102545-630 102545-632 102545-634
(2-Fluorophenyl)(2-(methylamino)-5-nitrophenyl)methanone ≥95%
(2-Fluorophenyl)(2-(methylamino)-5-nitrophenyl)methanone

Hazard: Irritant

Formula: C₁₄H₁₁FN₂O₃
MDL Number: MFCD06658167
CAS Number: 735-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR